| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016724.hg.1 |
ENST00000328942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016724.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016724.hg.1 |
NM_012375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016724.hg.1 |
TC11001326.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009912.hg.1 |
ENST00000328942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009912.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009912.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009912.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009912.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009912.hg.1 |
NM_012375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009912.hg.1 |
ENST00000328942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009912.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009912.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009912.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009912.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009912.hg.1 |
NM_012375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174825.hg.1 |
ENST00000328942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174825.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174825.hg.1 |
NM_012375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174825.hg.1 |
ENST00000328942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174825.hg.1 |
ENST00000380367.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174825.hg.1 |
NM_012375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174825.hg.1 |
TC1100009912.hg.1 |
Clariom D TC |