| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011047.hg.1 |
ENST00000368444.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011047.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011047.hg.1 |
TC06000949.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011048.hg.1 |
ENST00000368444.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011048.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011048.hg.1 |
TC06000949.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011049.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011049.hg.1 |
ENST00000368444.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011049.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011049.hg.1 |
TC06000949.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011050.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011050.hg.1 |
ENST00000368444.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011050.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011050.hg.1 |
TC06000949.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017988.hg.1 |
ENST00000424004.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163908.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163908.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163908.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163908.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163908.hg.1 |
TC0600009332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600163910.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163910.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163910.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163910.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163910.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163910.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163910.hg.1 |
TC0600009332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600163909.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163909.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163909.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163909.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163909.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163909.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163909.hg.1 |
TC0600009332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100017988.hg.1 |
ENST00000424004.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009332.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163907.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163907.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163907.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163907.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163907.hg.1 |
TC0600009332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600163911.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163911.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163911.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163911.hg.1 |
ENST00000356535.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163911.hg.1 |
ENST00000368444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600163911.hg.1 |
NM_001446.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600163911.hg.1 |
TC0600009332.hg.1 |
Clariom D TC |