| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
NM_003510_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124525 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8117601 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8124526 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8124524 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8117600 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
239041_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
214644_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11759219_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11715162_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11759220_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11715162_PM_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11759219_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11759220_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016350.hg.1 |
ENST00000330180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016350.hg.1 |
NM_003510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016350.hg.1 |
TC06001425.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000396980.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000396980.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000449538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000449538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000449538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000449538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000449538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000613174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
ENST00000629628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
NM_003511.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
NM_003511.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
NM_003511.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
NM_003520.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007380.hg.1 |
NM_003520.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016351.hg.1 |
ENST00000330180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016351.hg.1 |
NM_003510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016351.hg.1 |
TC06001425.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011228.hg.1 |
ENST00000618958.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011228.hg.1 |
NM_003510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011228.hg.1 |
NM_003510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175721.hg.1 |
ENST00000618958.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175721.hg.1 |
NM_003510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175721.hg.1 |
ENST00000618958.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175721.hg.1 |
NM_003510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175721.hg.1 |
TC0600011228.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600175720.hg.1 |
NM_003510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175720.hg.1 |
NM_003510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175720.hg.1 |
TC0600011228.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600175718.hg.1 |
TC0600011228.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600175722.hg.1 |
TC0600011228.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000396980.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000396980.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000449538.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000449538.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000449538.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000449538.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000449538.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000606613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000612898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000613174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
ENST00000629628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
NM_003511.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
NM_003511.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
NM_003511.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
NM_003520.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007380.hg.1 |
NM_003520.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011228.hg.1 |
ENST00000618958.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011228.hg.1 |
NM_003510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011228.hg.1 |
NM_003510.2 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
214644_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
214644_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
208583_PM_x_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
239041_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
239041_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
207156_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
214542_x_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
214554_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
214554_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
214542_PM_x_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
208583_x_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
207156_at |
- |
- |
| Human Genome U133A 2.0 Array |
214554_PM_at |
- |
- |
| Human Genome U133A 2.0 Array |
214542_PM_x_at |
- |
- |
| Human Genome U133A 2.0 Array |
214644_PM_at |
- |
- |
| Human Genome U133A 2.0 Array |
214554_at |
- |
- |
| Human Genome U133A 2.0 Array |
207156_at |
- |
- |
| Human Genome U133A 2.0 Array |
214542_x_at |
- |
- |
| Human Genome U133A 2.0 Array |
208583_PM_x_at |
- |
- |
| Human Genome U133A 2.0 Array |
214644_at |
- |
- |
| Human Genome U133A 2.0 Array |
208583_x_at |
- |
- |
| Human Genome U133A 2.0 Array |
207156_PM_at |
- |
- |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_003510_at |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw561891 |
- |
- |
| Human Genome U95 Set |
34156_i_at |
- |
- |
| Human Genome U95 Set |
31749_PM_f_at |
- |
- |
| Human Genome U95 Set |
285_PM_g_at |
- |
- |
| Human Genome U95 Set |
34157_f_at |
- |
- |
| Human Genome U95 Set |
31768_at |
- |
- |
| Human Genome U95 Set |
34156_PM_i_at |
- |
- |
| Human Genome U95 Set |
31749_f_at |
- |
- |
| Human Genome U95 Set |
34157_PM_f_at |
- |
- |
| Human Genome U95 Set |
284_at |
- |
- |
| Human Genome U95 Set |
284_PM_at |
- |
- |
| Human Genome U95 Set |
285_g_at |
- |
- |
| Human Genome U95 Set |
31768_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
214644_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239041_at9 |
- |
- |
| Human Genome U95 Set |
83824_at |
- |
- |
| Human Genome U95 Set |
83824_PM_at |
- |
- |
| Human Genome U133 Set |
208583_PM_x_at |
- |
- |
| Human Genome U133 Set |
214554_PM_at |
- |
- |
| Human Genome U133 Set |
214644_PM_at |
- |
- |
| Human Genome U133 Set |
207156_at |
- |
- |
| Human Genome U133 Set |
214542_PM_x_at |
- |
- |
| Human Genome U133 Set |
214644_at |
- |
- |
| Human Genome U133 Set |
214542_x_at |
- |
- |
| Human Genome U133 Set |
208583_x_at |
- |
- |
| Human Genome U133 Set |
214554_at |
- |
- |
| Human Genome U133 Set |
207156_PM_at |
- |
- |
| Human Genome U133 Set |
239041_at |
- |
- |
| Human Genome U133 Set |
239041_PM_at |
- |
- |