| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08003969.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08003969.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08003969.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08003969.hg.1 |
TC08000287.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000521311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000521311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000521311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
ENST00000521311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800150337.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800150337.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800150337.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800150337.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800150337.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800150337.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800150337.hg.1 |
TC0800007328.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000287322.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000432471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000521282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000521311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000521311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000521311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
ENST00000521311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_001204878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007328.hg.1 |
NM_004874.3 |
Refseq |