Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00365575_m1
          See other ASB16 GE Assays ›
          Gene Symbol:
          ASB16
          Gene Name:
          ankyrin repeat and SOCS box containing 16
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 44170706 - 44179083 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00365575_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ASB16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080863.4 NP_543139.4 2 - 3 650 79
          GenBank mRNA AF403034.1 - 1 - 2 265 79
          AK054727.1 - 2 - 3 650 79
          AK314595.1 - 2 - 3 642 79
          AY557346.1 - 2 - 3 566 79
          BC075088.2 - 2 - 3 599 30915388 79
          BC118584.1 - 2 - 3 632 40084995 79
          BC118585.1 - 2 - 3 632 40084992 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700007983.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000564.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ASB16

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          intracellular signal transduction
          ubiquitin-protein transferase activity
          protein binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17006868.hg.1 ENST00000293414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17006868.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17006868.hg.1 TC17000564.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000591700.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000591700.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 ENST00000591700.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17006867.hg.1 ENST00000293414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17006867.hg.1 NM_080863.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17006867.hg.1 TC17000564.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700158389.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158389.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158389.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700158389.hg.1 TC1700007983.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700158389.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158389.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158389.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700158390.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158390.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700158390.hg.1 TC1700007983.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700158390.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158390.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000591700.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000591700.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 ENST00000591700.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay TC1700007983.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700158388.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158388.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158388.hg.1 NM_080863.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700158388.hg.1 TC1700007983.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700158388.hg.1 ENST00000293414.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158388.hg.1 ENST00000589618.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700158388.hg.1 NM_080863.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline