Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00372674_g1
          See other GATAD2B GE Assays ›
          Gene Symbol:
          GATAD2B
          Gene Name:
          GATA zinc finger domain containing 2B
          Gene Aliases:
          MRD18, P66beta, p68
          Chromosome Location:
          Chr.1: 153804725 - 153922975 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00372674_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GATAD2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005245364.4 XP_005245421.1 9 - 10 1628 101
          GenBank mRNA AB032976.1 - 8 - 9 1253 101
          AF411836.1 - 9 - 10 1550 101
          AK094665.1 - 9 - 10 1777 101
          BC069419.1 - 9 - 10 1550 7262291 101
          BC112052.1 - 9 - 10 1554 8327520 101
          BC112080.1 - 9 - 10 1554 8327548 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100015885.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA methylation
          transcription, DNA-templated
          ATP-dependent chromatin remodeling
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          zinc ion binding
          nucleosomal DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015885.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219357.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219357.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219357.hg.1 ENST00000368655.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219357.hg.1 NM_020699.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219357.hg.1 TC0100015885.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline