Search
Search

ARHGEF6| Gene Symbol: ARHGEF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306177.1 | NP_001293106.1 | 13 - 14 | 1555 | 90 | |
| NM_004840.2 | NP_004831.1 | 14 - 15 | 2017 | 90 | ||
| XM_005262499.2 | XP_005262556.1 | 14 - 15 | 2329 | 90 | ||
| XM_011531412.2 | XP_011529714.1 | 15 - 16 | 2410 | 90 | ||
| XM_011531413.1 | XP_011529715.1 | 14 - 15 | 2329 | 90 | ||
| XM_011531414.1 | XP_011529716.1 | 14 - 15 | 2329 | 90 | ||
| XM_011531415.2 | XP_011529717.1 | 14 - 15 | 1765 | 90 | ||
| XM_011531416.2 | XP_011529718.1 | 14 - 15 | 2028 | 90 | ||
| XM_011531417.1 | XP_011529719.1 | 13 - 14 | 1947 | 90 | ||
| XM_017029975.1 | XP_016885464.1 | 15 - 16 | 2410 | 90 | ||
| XM_017029976.1 | XP_016885465.1 | 13 - 14 | 1947 | 90 | ||
| GenBank mRNA | AK291742.1 | - | 14 - 15 | 1560 | 90 | |
| AK294929.1 | - | 13 - 14 | 1361 | 90 | ||
| AK295709.1 | - | 14 - 15 | 1637 | 90 | ||
| BC033768.1 | - | 2 - 3 | 188 | 90 | ||
| BC039856.1 | - | 14 - 15 | 2017 | 4510206 | 90 | |
| BC043505.1 | - | 13 - 14 | 1521 | 6015396 | 90 | |
| BX537390.1 | - | 14 - 15 | 1622 | 90 | ||
| D13631.1 | - | 6 - 7 | 595 | 90 | ||
| D25304.1 | - | 14 - 15 | 1548 | 90 | ||
Set Membership: |
|