| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000521738.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000521738.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000521738.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000521738.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
ENST00000521738.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05012801.hg.1 |
ENST00000296786.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05012801.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05012801.hg.1 |
TC05000886.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500164713.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500164713.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500164713.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500164713.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500164713.hg.1 |
TC0500009250.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000296786.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000519276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000521738.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000521738.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000521738.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000521738.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
ENST00000521738.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009250.hg.1 |
NM_145049.4 |
Refseq |