| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000475775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000475775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000475775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000602308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000602308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000602308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000602308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000602308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000602582.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000602582.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000602582.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000602582.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000602582.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
ENST00000622338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017612.hg.1 |
NR_106800.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000284551.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000366699.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000475775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000475775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000475775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000493030.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000602308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000602308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000602308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000602308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000602308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000602582.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000602582.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000602582.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000602582.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000602582.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
ENST00000622338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NM_145214.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017612.hg.1 |
NR_106800.1 |
Refseq |