| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8045859 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8045858 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1555224_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02013969.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02013969.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02013969.hg.1 |
TC02000959.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02013970.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02013970.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02013970.hg.1 |
TC02000959.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009752.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009752.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009752.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009752.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009752.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009752.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009752.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009752.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009752.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009752.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168038.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200168038.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168038.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200168038.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168038.hg.1 |
TC0200009752.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200009752.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009752.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009752.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009752.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009752.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009752.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009752.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009752.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009752.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009752.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168039.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200168039.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168039.hg.1 |
ENST00000412781.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200168039.hg.1 |
NR_038850.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168039.hg.1 |
TC0200009752.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1555224_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1555224_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555224_at11 |
- |
- |