| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013349.hg.1 |
ENST00000454181.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013349.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013349.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15013349.hg.1 |
TC15001444.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000560491.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000560491.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000560491.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000560491.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
ENST00000560491.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500169362.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169362.hg.1 |
TC1500009438.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500169362.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000267838.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000560491.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000560491.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000560491.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000560491.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
ENST00000560491.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009438.hg.1 |
NM_153374.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500169360.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169360.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169360.hg.1 |
NM_001143917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500169360.hg.1 |
TC1500009438.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500169360.hg.1 |
ENST00000454181.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169360.hg.1 |
ENST00000558126.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500169360.hg.1 |
NM_001143917.1 |
Refseq |