Search
Search

WDR33| Gene Symbol: WDR33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018383.4 | NP_060853.3 | 20 - 21 | 3767 | 138 | |
| XM_005263697.2 | XP_005263754.1 | 19 - 20 | 3328 | 138 | ||
| XM_011511436.1 | XP_011509738.1 | 20 - 21 | 3796 | 138 | ||
| GenBank mRNA | AB044749.1 | - | 20 - 21 | 3727 | 138 | |
| AK098003.1 | - | 5 - 6 | 138 | |||
| BC010283.1 | - | 5 - 6 | 1112 | 138 | ||
Set Membership: |
|