Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00384755_m1
          See other ATP23 GE Assays ›
          Gene Symbol:
          ATP23
          Gene Name:
          ATP23 metallopeptidase and ATP synthase assembly factor homolog
          Gene Aliases:
          KUB3, XRCC6BP1
          Chromosome Location:
          Chr.12: 57941504 - 57957269 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00384755_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATP23
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320408.1 NP_001307337.1 4 - 5 690 68
          NM_001320409.1 NP_001307338.1 3 - 4 604 68
          NM_001320410.1 NP_001307339.1 5 - 6 821 68
          NM_033276.3 NP_150592.1 4 - 5 650 68
          XM_011538976.2 XP_011537278.1 3 - 4 325 68
          XM_017020206.1 XP_016875695.1 4 - 5 440 68
          GenBank mRNA AF078164.2 - 4 - 5 580 68
          BC012776.2 - 4 - 5 492 3936722 68
          BC115381.1 - 4 - 5 491 40036852 68
          BC115382.1 - 4 - 5 490 40036859 68
          BM544500.1 - 4 - 5 649 68
          BM557436.1 - 5 - 6 655 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200007906.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000547.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ATP23

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via nonhomologous end joining
          protein phosphorylation
          proteolysis
          metalloendopeptidase activity
          DNA-dependent protein kinase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12007369.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12007369.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12007369.hg.1 TC12000547.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12007370.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12007370.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12007370.hg.1 TC12000547.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000549257.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000549257.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000549257.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000549257.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200158029.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158029.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200158029.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158029.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200158029.hg.1 TC1200007906.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200158030.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158030.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200158030.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158030.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200158030.hg.1 TC1200007906.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200158027.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158027.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158027.hg.1 ENST00000549257.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158027.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200158027.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158027.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158027.hg.1 ENST00000549257.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158027.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200158027.hg.1 TC1200007906.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000549257.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000549257.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000549257.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000549257.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 ENST00000551997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay TC1200007906.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200158028.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158028.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158028.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200158028.hg.1 ENST00000300145.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158028.hg.1 ENST00000546709.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200158028.hg.1 NM_033276.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200158028.hg.1 TC1200007906.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 177472 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline