| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036292.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036292.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036292.hg.1 |
TC01002352.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013305.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198731.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198731.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198731.hg.1 |
ENST00000374479.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198731.hg.1 |
NM_000147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198731.hg.1 |
TC0100013305.hg.1 |
Clariom D TC |