Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00398731_m1
          See other FAAH2 GE Assays ›
          Gene Symbol:
          FAAH2
          Gene Name:
          fatty acid amide hydrolase 2
          Gene Aliases:
          AMDD
          Chromosome Location:
          Chr.X: 57121861 - 57489196 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00398731_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FAAH2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_174912.3 NP_777572.2 3 - 4 563 105
          XM_005261982.1 XP_005262039.1 3 - 4 662 105
          XM_005261985.2 XP_005262042.1 4 - 5 425 105
          XM_005261986.3 XP_005262043.1 3 - 4 662 105
          XM_005261987.3 XP_005262044.1 3 - 4 662 105
          XM_011530767.2 XP_011529069.1 4 - 5 503 105
          XM_017029290.1 XP_016884779.1 3 - 4 662 105
          XM_017029293.1 XP_016884782.1 3 - 4 662 105
          XM_017029294.1 XP_016884783.1 3 - 4 662 105
          GenBank mRNA AK055766.1 - 3 - 4 534 105
          BC048279.1 - 3 - 4 351 105
          BC073922.1 - 3 - 4 529 5187808 105
          EU285560.1 - 3 - 4 414 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00007416.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000346.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FAAH2

          Panther Classification:

          Molecular Function -

          hydrolase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          arachidonic acid metabolic process
          carbon-nitrogen ligase activity, with glutamine as amido-N-donor
          fatty acid amide hydrolase activity
          anandamide amidohydrolase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000374900.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000465623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000465623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000465623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000465623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000465623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000491179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000491179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000491179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 ENST00000491179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007416.hg.1 NM_174912.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline