| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015291.hg.1 |
ENST00000221480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015291.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015291.hg.1 |
TC19001113.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009479.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176224.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176224.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176224.hg.1 |
TC1900009479.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900176224.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176224.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
NM_080662.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
TC1900009479.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
ENST00000221480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
ENST00000593547.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
ENST00000593942.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
ENST00000599519.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
NM_001270539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
NM_001300881.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176217.hg.1 |
NM_080662.3 |
Refseq |