| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116853 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116789 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116846 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116870 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116804 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116843 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116844 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624684 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116871 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116879 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116809 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116778 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624685 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624663 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786411 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786375 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116848 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624674 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116761 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624660 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786455 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116763 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624677 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624704 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624655 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116910 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786468 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624675 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116790 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786417 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624701 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786474 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116893 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116787 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624696 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786479 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624656 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116788 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116786 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116851 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116832 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786428 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786460 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116770 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116924 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624653 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116773 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116850 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116757 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786473 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786446 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116839 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116849 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116842 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116794 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116836 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624695 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786387 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116878 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116813 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116768 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786477 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16786433 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30116807 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624678 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09624657 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
100678 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
98913 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
33636 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 8x60K |
26179 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
80419 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
545927 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
865205 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
801337 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
87242 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
451470 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
78345 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
770647 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
857305 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
214845 |
AJ412023.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
214845 |
AJ412024.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
214845 |
AK095311.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
214845 |
BC004131.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
214845 |
BC020692.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
214845 |
CR600970.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
214845 |
CR605740.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
214845 |
Y15227.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
418324 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
245827 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
115190 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
144507 |
XM_001130910.1 |
refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
297956 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
173028 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
68472 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
135776 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
278297 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P325593 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P325593 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
6492 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P116290 |
- |
- |
| Agilent Human Genome CGH Microarray 44A |
A_14_P116290 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
3939 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P20233665 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P03019774 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P20233711 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_18_P12206785 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P33684032 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P20233677 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P20233674 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P325593 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_23_P325593 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16786375 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624651 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116759 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116770 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116778 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116761 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624655 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624656 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116762 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116768 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624649 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116763 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624657 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116773 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624648 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624660 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116760 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16786369 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624649 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116762 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116770 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116760 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116759 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116773 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624648 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624657 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624656 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16786369 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116763 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116768 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624660 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624651 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16786375 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116761 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30116778 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09624655 |
- |
- |
| Whole Human Genome Oligo Microarray G4112A |
A_23_P325593 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P116290 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09624652 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09624648 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09624659 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09624656 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09624661 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09624650 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09624647 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09624655 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P03019762 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P20233674 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P116290 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P03019774 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P116290 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P325593 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
119815 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
42089 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
101498 |
CR621082.1 |
genbank_mrna_human |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03019774 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P116290 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03019760 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03019758 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P40383508 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03019762 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P12204247 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P12201753 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P20233677 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P12204121 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P20233710 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P20233674 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624647 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624657 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624661 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624652 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624683 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624678 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16786477 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624687 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624681 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624664 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624650 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624670 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624691 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16786379 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624676 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624668 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624700 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624699 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624701 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624674 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624704 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624690 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624666 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624696 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624685 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16786369 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624672 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624695 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624655 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624693 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09624659 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P03019762 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P20233674 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P03019774 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P116290 |
- |
- |