| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025298.hg.1 |
ENST00000515823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025298.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025298.hg.1 |
TC05003431.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013372.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500196899.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500196899.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500196899.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500196899.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500196899.hg.1 |
ENST00000618155.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500196899.hg.1 |
NM_001077693.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500196899.hg.1 |
NM_001293739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500196899.hg.1 |
NR_121659.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500196899.hg.1 |
TC0500013372.hg.1 |
Clariom D TC |