Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00416910_m1
          See other TMEM240 GE Assays ›
          Gene Symbol:
          TMEM240
          Gene Name:
          transmembrane protein 240
          Gene Aliases:
          C1orf70, SCA21
          Chromosome Location:
          Chr.1: 1534778 - 1540360 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00416910_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TMEM240
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001114748.1 NP_001108220.1 2 - 3 176 153
          GenBank mRNA BC147018.1 - 3 - 4 205 9056946 153
          BC147020.1 - 3 - 4 205 9056948 153
          BU929336.1 - 2 - 3 157 153

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100012485.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002101.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20006814 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001827 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001831 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15009587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20006832 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001839 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001832 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20006838 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001836 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001834 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20006852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20006843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15009602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20006830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15009588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15009600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15009597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20006844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20006842 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 100397 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 778128 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 320198 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 134348 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3219459 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P161913 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 45001 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2297 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 41354 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00000687 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P161913 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3219459 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P161913 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3219459 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20006843 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20006842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001839 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001840 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20006832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20006852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001836 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001845 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20006844 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001840 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20006844 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001836 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20006852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001845 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20006832 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20006842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20006843 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00001839 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P161913 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P17423492 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00000687 - -
          Agilent Human 1B Microarray G4111A A_32_P66424 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001825 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15009591 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001840 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001826 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001833 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15009522 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001810 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001821 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001842 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001839 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001824 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001820 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15009611 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001844 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15009592 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001845 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001837 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001847 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15009588 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15009584 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001830 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001846 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15009597 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001827 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15009602 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15009604 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00000687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Genome U219 Array 11737469_PM_at - -
          Affymetrix Human Genome U219 Array 11737469_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000624426.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000624426.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 ENST00000624426.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01032053.hg.1 ENST00000378733.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01032053.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01032053.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01032053.hg.1 TC01002101.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01032055.hg.1 ENST00000378733.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01032055.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01032055.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01032055.hg.1 TC01002101.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01032054.hg.1 ENST00000378733.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01032054.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01032054.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01032054.hg.1 TC01002101.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100192084.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192084.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192084.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100192084.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192084.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192084.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100192084.hg.1 TC0100012485.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100192092.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192092.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192092.hg.1 TC0100012485.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100192086.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192086.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192086.hg.1 ENST00000624426.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192086.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100192086.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192086.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192086.hg.1 ENST00000624426.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192086.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100192086.hg.1 TC0100012485.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000624426.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000624426.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 ENST00000624426.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay TC0100012485.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100192087.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192087.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192087.hg.1 ENST00000624426.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192087.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100192087.hg.1 ENST00000378733.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192087.hg.1 ENST00000425828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192087.hg.1 ENST00000624426.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192087.hg.1 NM_001114748.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100192087.hg.1 TC0100012485.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A042135 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A042135 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 152752 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1748257 NM_001114748.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1748257 BC147018.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1748257 BC147020.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1748257 BU929336.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1751459 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1748257 NM_001114748.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1748257 BC147018.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1748257 BC147020.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1748257 BU929336.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1748257 NM_001114748.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1748257 BC147018.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1748257 BC147020.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1748257 BU929336.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline