Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00417354_m1
          See other LINC00272 GE Assays ›
          Gene Symbol:
          LINC00272
          Gene Name:
          long intergenic non-protein coding RNA 272
          Gene Aliases:
          C1orf120, NCRNA00272
          Chromosome Location:
          Chr.1: 182407621 - 182414813 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00417354_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00272
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034131.1 3 - 4 610 71
          GenBank mRNA AF508909.1 - 3 - 4 610 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010843.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001577.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15149827 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702297 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702293 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702272 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702265 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15149821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642613 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15149828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702274 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15149825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702301 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702267 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642631 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702288 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702266 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15149826 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642614 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702280 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00642608 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20702261 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20124 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3412980 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 155981 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15365543 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00205855 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15365537 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00205863 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7907974 - -
          Affymetrix Human Gene 1.0 ST Array 7907973 - -
          Affymetrix Human Gene 1.0 ST Array 7907975 - -
          Affymetrix Human Gene 1.0 ST Array 7907976 - -
          Affymetrix Human Gene 1.1 ST Array 7907972 - -
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000612603.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000612603.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 ENST00000612603.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01024330.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01024330.hg.1 NR_034131.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01024330.hg.1 TC01001577.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01024331.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01024331.hg.1 NR_034131.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01024331.hg.1 TC01001577.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100179645.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100179645.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100179645.hg.1 NR_034131.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100179645.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100179645.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100179645.hg.1 NR_034131.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100179645.hg.1 TC0100010843.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000612603.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000612603.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 ENST00000612603.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010843.hg.1 NR_034131.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100179646.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100179646.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100179646.hg.1 NR_034131.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100179646.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100179646.hg.1 ENST00000367563.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100179646.hg.1 NR_034131.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100179646.hg.1 TC0100010843.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036181 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036181 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 102441 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1755835 NR_034131.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1755835 AF508909.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1755835 NR_034131.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1755835 AF508909.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1755835 NR_034131.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1755835 AF508909.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1755835 NR_034131.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1755835 AF508909.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1755835 NR_034131.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1755835 AF508909.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline