Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00426397_m1
          See other CYP2C9 GE Assays ›
          Gene Symbol:
          CYP2C9
          Gene Name:
          cytochrome P450 family 2 subfamily C member 9
          Gene Aliases:
          CPC9, CYP2C, CYP2C10, CYPIIC9, P450IIC9
          Chromosome Location:
          Chr.10: 94905867 - 94989395 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00426397_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CYP2C9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000771.3 NP_000762.2 6 - 7 986 148
          XM_017015758.1 XP_016871247.1 7 - 8 1406 148
          GenBank mRNA AK289420.1 - 6 - 7 986 148
          AK298458.1 - 6 - 7 986 148
          BC125054.1 - 6 - 7 961 40116637 148
          D00173.1 - 6 - 7 952 148
          M15331.1 - 6 - 7 946 148
          M21939.1 - 6 - 7 947 148
          M21940.1 - 4 - 5 640 148
          M61855.1 - 6 - 7 973 148
          M61857.1 - 6 - 7 971 148
          S46963.1 - 6 - 7 924 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008508.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000675.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection P450 Homo sapiens Human CYP450 and other Oxygenases Inventoried MicroArray Quality Control (MAQC) Human CYP450 + oxygenases Human_CYP450_and_other_Oxygenases Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CYP2C9

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          steroid metabolic process
          monoterpenoid metabolic process
          drug metabolic process
          epoxygenase P450 pathway
          urea metabolic process
          monocarboxylic acid metabolic process
          drug catabolic process
          exogenous drug catabolic process
          cellular amide metabolic process
          oxidation-reduction process
          oxidative demethylation
          omega-hydroxylase P450 pathway
          cholesterol 25-hydroxylase activity
          monooxygenase activity
          iron ion binding
          drug binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity
          (S)-limonene 6-monooxygenase activity
          (S)-limonene 7-monooxygenase activity
          oxygen binding
          heme binding
          caffeine oxidase activity
          (R)-limonene 6-monooxygenase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10007741.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10007741.hg.1 ENST00000545448.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10007741.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10007741.hg.1 TC10000675.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000480405.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000480405.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000480405.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000480405.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 ENST00000480405.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10007742.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10007742.hg.1 ENST00000545448.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10007742.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10007742.hg.1 TC10000675.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000461906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000461906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000461906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000461906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000461906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000473496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000473496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000473496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000473496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 ENST00000473496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10007744.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10007744.hg.1 NM_000771.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10007744.hg.1 TC10000675.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000371321.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000480405.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000480405.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000480405.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000480405.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 ENST00000480405.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay TC1000012481.hg.1 NM_000769.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000158002.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000158002.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000158002.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000158002.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000158002.hg.1 TC1000008508.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000158004.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000158004.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000158004.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000158004.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000158004.hg.1 TC1000008508.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000158003.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000158003.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000158003.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000158003.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000158003.hg.1 TC1000008508.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000260682.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000461906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000461906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000461906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000461906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000461906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000473496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000473496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000473496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000473496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 ENST00000473496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008508.hg.1 NM_000771.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline