Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00426835_g1
          See other ACTA2 GE Assays ›
          Gene Symbol:
          ACTA2
          Gene Name:
          actin alpha 2, smooth muscle
          Gene Aliases:
          ACTSA, SMDYS
          Chromosome Location:
          Chr.10: 88935074 - 88991337 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00426835_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ACTA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001141945.3 NP_001135417.1 2 - 3 553 105
          NM_001320855.2 NP_001307784.1 2 - 3 470 105
          NM_001406462.1 NP_001393391.1 3 - 4 628 105
          NM_001406463.1 NP_001393392.1 3 - 4 711 105
          NM_001406464.1 NP_001393393.1 3 - 4 342 105
          NM_001406466.1 NP_001393395.1 2 - 3 197 105
          NM_001406471.1 NP_001393400.1 2 - 3 553 105
          NM_001613.4 NP_001604.1 2 - 3 197 105
          GenBank mRNA AK300353.1 - 2 - 3 196 105
          AK313294.1 - 2 - 3 197 105
          AY692464.1 - 2 - 3 523 105
          BC017554.2 - 2 - 3 185 105
          BC093052.1 - 2 - 3 196 105
          BP248338.1 - 2 - 3 459 105
          BP320140.1 - 2 - 3 301 105
          BP379410.1 - 2 - 3 433 105
          CR536518.1 - 1 - 2 131 105
          DC385559.1 - 2 - 3 362 105
          J05192.1 - 2 - 3 179 105
          X13839.1 - 2 - 3 178 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000011370.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001498.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ACTA2

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          regulation of blood pressure
          response to virus
          positive regulation of gene expression
          vascular smooth muscle contraction
          positive regulation of hepatic stellate cell migration
          positive regulation of hepatic stellate cell contraction
          positive regulation of ERK1 and ERK2 cascade
          cellular response to transforming growth factor beta stimulus
          juxtaglomerular apparatus development
          glomerular mesangial cell development
          mesenchyme migration
          positive regulation of hepatic stellate cell activation
          nucleotide binding
          protein binding
          ATP binding
          hydrolase activity
          protein kinase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000224784.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000224784.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000224784.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000224784.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000224784.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000224784.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000224784.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000224784.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000224784.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000415557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000415557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000415557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000415557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000415557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000415557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000415557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458159.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458159.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458159.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458159.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458159.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000458208.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000480297.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000480297.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000480297.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000480297.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000480297.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000480297.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000482085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000482085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000488967.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000488967.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 ENST00000488967.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001141945.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001613.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001613.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001613.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001613.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001613.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001613.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001613.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001613.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001613.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011370.hg.1 NM_001613.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 201689 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline