Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00430304_g1
          See other DDIT4 GE Assays ›
          Gene Symbol:
          DDIT4
          Gene Name:
          DNA damage inducible transcript 4
          Gene Aliases:
          Dig2, REDD-1, REDD1
          Chromosome Location:
          Chr.10: 72273924 - 72276036 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00430304_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DDIT4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019058.4 NP_061931.1 1 - 2 141 82
          GenBank mRNA AF335324.2 - 1 - 2 130 82
          AK000507.1 - 1 - 2 141 82
          AL136668.1 - 1 - 2 145 82
          BC007714.1 - 1 - 2 134 82
          BC015236.1 - 1 - 2 120 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000007990.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000449.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AL136668.1, BC007714.1, BC015236.1, NM_019058.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Human Hypoxia Panel 1700 Inventoried Human_Hypoxia Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DDIT4

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neuron migration
          apoptotic process
          brain development
          negative regulation of signal transduction
          neuron differentiation
          negative regulation of TOR signaling
          protein-containing complex disassembly
          intracellular signal transduction
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          negative regulation of glycolytic process
          neurotrophin TRK receptor signaling pathway
          defense response to virus
          cellular response to dexamethasone stimulus
          reactive oxygen species metabolic process
          protein binding
          14-3-3 protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10005308.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10005308.hg.1 NM_019058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10005308.hg.1 TC10000449.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10005310.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10005310.hg.1 NM_019058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10005310.hg.1 TC10000449.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10005312.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10005312.hg.1 NM_019058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10005312.hg.1 TC10000449.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000154195.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154195.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154195.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154195.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000154195.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154195.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154195.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154195.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000154195.hg.1 TC1000007990.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay TC1000007990.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000154190.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154190.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154190.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000154190.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154190.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154190.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000154190.hg.1 TC1000007990.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000154194.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154194.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154194.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154194.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000154194.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154194.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154194.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154194.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000154194.hg.1 TC1000007990.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000154192.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154192.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154192.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154192.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000154192.hg.1 ENST00000307365.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154192.hg.1 ENST00000471240.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154192.hg.1 ENST00000473155.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154192.hg.1 NM_019058.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000154192.hg.1 TC1000007990.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 160257 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline