Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00535896_s1
          See other PPP1R2P9 GE Assays ›
          Gene Symbol:
          PPP1R2P9
          Gene Name:
          protein phosphatase 1 regulatory inhibitor subunit 2 pseudogene 9
          Gene Aliases:
          I-4
          Chromosome Location:
          Chr.X: 42777366 - 42778235 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00535896_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PPP1R2P9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002191.2 1 151 106
          GenBank mRNA AB044137.1 - 1 151 106
          BC056673.1 - 1 81 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00009475.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000976.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AB044137.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator phosphatase inhibitor phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of signal transduction
          negative regulation of catalytic activity
          regulation of phosphoprotein phosphatase activity
          protein phosphatase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17337197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17337199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723021 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428894 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723044 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428839 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428889 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723048 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11723043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32428896 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P34007 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P34007 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 6074 - -
          Agilent Human Genome CGH Microarray 44B A_14_P122947 - -
          Agilent Human Genome CGH Microarray 44A A_14_P122947 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 44710 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P34007 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P34007 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428875 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428869 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428879 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428890 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428880 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428891 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428890 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428869 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428880 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428875 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428879 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11723046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32428891 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P34007 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P122947 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P122947 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P122947 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P34007 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 41128 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 96201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723060 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723021 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723010 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723007 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17337195 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723025 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723004 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723037 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723001 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17337199 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723047 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723040 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723008 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723006 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723000 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723039 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723013 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723055 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723028 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723046 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723017 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723050 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723019 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723015 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723024 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723043 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723034 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723058 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11722999 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723031 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723048 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723044 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11723009 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002191_at - -
          Affymetrix Human Gene 1.0 ST Array 8172198 - -
          Affymetrix Human Gene 1.0 ST Array 8172200 - -
          Affymetrix Human Gene 1.0 ST Array 8172201 - -
          Affymetrix Human Gene 1.0 ST Array 8172199 - -
          Affymetrix Human Gene 1.1 ST Array 8172197 - -
          Affymetrix HT HG-U133+ PM Array Plate 207377_PM_at - -
          Affymetrix Human Genome U219 Array 11745031_PM_at - -
          Affymetrix Human Genome U219 Array 11745031_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0X00009475.hg.1 ENST00000378131.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009475.hg.1 NR_002191.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009475.hg.1 NR_002191.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009475.hg.1 NR_002191.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X013005.hg.1 NR_002191.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X013005.hg.1 TC0X000976.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00164758.hg.1 NR_002191.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00164758.hg.1 NR_002191.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00164758.hg.1 TC0X00009475.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00164757.hg.1 ENST00000378131.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00164757.hg.1 NR_002191.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00164757.hg.1 ENST00000378131.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00164757.hg.1 NR_002191.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00164757.hg.1 TC0X00009475.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0X00009475.hg.1 ENST00000378131.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009475.hg.1 NR_002191.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009475.hg.1 NR_002191.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009475.hg.1 NR_002191.2 Refseq
          Human Genome U133 Plus 2.0 Array 207377_at - -
          Human Genome U133 Plus 2.0 Array 207377_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_002191_at - -
          AffymetrixPathwork Diagnostics Pathchip pw543087 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207377_at4 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A039775 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A039774 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A039775 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A039774 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 190425 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2204142 AB044137.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2204142 BC056673.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2204142 NR_002191.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1766057 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2204142 AB044137.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2204142 BC056673.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2204142 NR_002191.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2204142 AB044137.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2204142 BC056673.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2204142 NR_002191.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2204142 AB044137.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2204142 BC056673.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2204142 NR_002191.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1766057 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_5701 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_5701 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2204142 AB044137.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2204142 BC056673.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2204142 NR_002191.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2204142 AB044137.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2204142 BC056673.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2204142 NR_002191.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2204142 AB044137.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2204142 BC056673.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2204142 NR_002191.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1766057 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2204142 AB044137.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2204142 BC056673.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2204142 NR_002191.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline