Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00540164_s1
          See other KRTAP1-5 GE Assays ›
          Gene Symbol:
          KRTAP1-5
          Gene Name:
          keratin associated protein 1-5
          Gene Aliases:
          KAP1.5, KRTAP1.5
          Chromosome Location:
          Chr.17: 41026026 - 41027202 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00540164_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KRTAP1-5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031957.1 NP_114163.1 1 561 105
          GenBank mRNA AJ406928.1 - 1 561 105
          BC069533.1 - 1 561 7262171 105
          BC093841.1 - 1 587 7939686 105
          BC101555.1 - 1 587 8069061 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010648.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001479.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KRTAP1-5

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 459389 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P118842 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3254579 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P118842 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 3094 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 12542 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42743 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3254579 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P118842 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3254579 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P118842 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P118842 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P138964 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P118842 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 8603 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_031957_at - -
          Affymetrix Human Gene 1.0 ST Array 8015184 - -
          Affymetrix Human Gene 1.0 ST Array 8019583 - -
          Affymetrix Human Gene 1.0 ST Array 8019590 - -
          Affymetrix Human Gene 1.0 ST Array 8019581 - -
          Affymetrix Human Gene 1.0 ST Array 8019582 - -
          Affymetrix Human Gene 1.0 ST Array 8019591 - -
          Affymetrix Human Gene 1.0 ST Array 8015199 - -
          Affymetrix Human Gene 1.0 ST Array 8015200 - -
          Affymetrix Human Gene 1.0 ST Array 8015192 - -
          Affymetrix Human Gene 1.0 ST Array 8019592 - -
          Affymetrix Human Gene 1.0 ST Array 8015203 - -
          Affymetrix Human Gene 1.0 ST Array 8015180 - -
          Affymetrix Human Gene 1.0 ST Array 8015185 - -
          Affymetrix Human Gene 1.0 ST Array 8015182 - -
          Affymetrix Human Gene 1.0 ST Array 8015186 - -
          Affymetrix Human Gene 1.0 ST Array 8019586 - -
          Affymetrix Human Gene 1.0 ST Array 8019589 - -
          Affymetrix Human Gene 1.0 ST Array 8015181 - -
          Affymetrix Human Gene 1.0 ST Array 8015183 - -
          Affymetrix Human Gene 1.1 ST Array 8015179 - -
          Affymetrix Human Gene 1.1 ST Array 8019588 - -
          Affymetrix HT HG-U133+ PM Array Plate 233533_PM_at - -
          Affymetrix Human Genome U219 Array 11729095_PM_at - -
          Affymetrix Human Genome U219 Array 11729095_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1700010648.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010648.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010648.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010648.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010648.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010648.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010648.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010648.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010648.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010648.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019661.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019661.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019661.hg.1 TC17001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019662.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019662.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019662.hg.1 TC17001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019666.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019666.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019666.hg.1 TC17001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019667.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019667.hg.1 ENST00000543389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019667.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019667.hg.1 TC17001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019664.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019664.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019664.hg.1 TC17001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019665.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019665.hg.1 ENST00000543389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019665.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019665.hg.1 TC17001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019668.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019668.hg.1 NM_031957.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019668.hg.1 TC17001479.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700184094.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184094.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184094.hg.1 TC1700010648.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184094.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184094.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184095.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184095.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184095.hg.1 TC1700010648.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184095.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184095.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184098.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184098.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184098.hg.1 TC1700010648.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184098.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184098.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010648.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010648.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010648.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010648.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010648.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010648.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010648.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010648.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010648.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010648.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184100.hg.1 TC1700010648.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184097.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184097.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184097.hg.1 TC1700010648.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184097.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184097.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184099.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184099.hg.1 NM_031957.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184099.hg.1 TC1700010648.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184099.hg.1 ENST00000361883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184099.hg.1 NM_031957.1 Refseq
          Human Genome U133 Plus 2.0 Array 233533_at - -
          Human Genome U133 Plus 2.0 Array 233533_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233533_at5 - -
          Human Genome U133 Set 233533_at - -
          Human Genome U133 Set 233533_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008306 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008306 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 183564 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1707652 NM_031957.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1707652 AJ406928.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1707652 BC069533.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1707652 BC093841.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1707652 BC101555.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1707652 NM_031957.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1707652 AJ406928.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1707652 BC069533.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1707652 BC093841.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1707652 BC101555.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1707652 NM_031957.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1707652 AJ406928.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1707652 BC069533.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1707652 BC093841.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1707652 BC101555.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_13994359-S NM_031957.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_13994359-S AJ406928.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_1272-S - -
          Illumina human-6 v1.0 expression beadchip 0006900440 - -
          Illumina humanRef-8 v1.0 expression beadchip 0006900440 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1707652 NM_031957.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1707652 AJ406928.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1707652 BC069533.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1707652 BC093841.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1707652 BC101555.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1707652 NM_031957.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1707652 AJ406928.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1707652 BC069533.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1707652 BC093841.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1707652 BC101555.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 3140544 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_1272 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1707652 NM_031957.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1707652 AJ406928.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1707652 BC069533.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1707652 BC093841.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1707652 BC101555.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1707652 NM_031957.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1707652 AJ406928.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1707652 BC069533.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1707652 BC093841.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1707652 BC101555.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1707652 NM_031957.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1707652 AJ406928.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1707652 BC069533.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1707652 BC093841.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1707652 BC101555.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline