Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00541043_g1
          See other SHMT1 GE Assays ›
          Gene Symbol:
          SHMT1
          Gene Name:
          serine hydroxymethyltransferase 1
          Gene Aliases:
          CSHMT, SHMT
          Chromosome Location:
          Chr.17: 18327860 - 18363563 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00541043_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SHMT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001281786.1 NP_001268715.1 5 - 6 690 65
          NM_004169.4 NP_004160.3 6 - 7 806 65
          NM_148918.2 NP_683718.1 6 - 7 806 65
          XM_005256767.3 XP_005256824.1 7 - 8 877 65
          XM_011523992.2 XP_011522294.1 6 - 7 774 65
          XM_017024957.1 XP_016880446.1 6 - 7 631 65
          XM_017024958.1 XP_016880447.1 7 - 8 877 65
          XM_017024959.1 XP_016880448.1 4 - 5 498 65
          XM_017024960.1 XP_016880449.1 4 - 5 498 65
          GenBank mRNA AK223552.1 - 6 - 7 737 65
          AK298415.1 - 5 - 6 622 65
          BC007979.2 - 6 - 7 652 3687873 65
          BC022874.1 - 6 - 7 656 4108106 65
          BC038598.1 - 6 - 7 663 4523709 65
          L11931.1 - 5 - 6 619 65
          L23928.1 - 5 - 6 607 65
          Y14485.1 - 6 - 7 772 65
          Y14486.1 - 1 - 2 917 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700009974.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001201.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SHMT1

          Panther Classification:

          Molecular Function -

          methyltransferase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          dTMP biosynthetic process
          glycine metabolic process
          L-serine metabolic process
          L-serine catabolic process
          purine nucleobase biosynthetic process
          glycine biosynthetic process from serine
          tetrahydrofolate interconversion
          carnitine biosynthetic process
          tetrahydrofolate metabolic process
          folic acid metabolic process
          protein tetramerization
          protein homotetramerization
          cellular response to tetrahydrofolate
          glycine hydroxymethyltransferase activity
          protein binding
          L-allo-threonine aldolase activity
          amino acid binding
          pyridoxal phosphate binding
          protein homodimerization activity
          serine binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000583780.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000583780.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000583780.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000583780.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000617912.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000618902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000618902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000618902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000618902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NR_106836.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline