| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22005677.hg.1 |
ENST00000216034.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22005677.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22005677.hg.1 |
TC22000303.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011872.hg.1 |
ENST00000528959.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008567.hg.1 |
ENST00000498134.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22005679.hg.1 |
ENST00000216034.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22005679.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22005679.hg.1 |
TC22000303.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008567.hg.1 |
ENST00000498134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200151831.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200151831.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200151831.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200151831.hg.1 |
TC2200007361.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200151831.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200151831.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200151831.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
ENST00000492561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007361.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011872.hg.1 |
ENST00000528959.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200151829.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200151829.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200151829.hg.1 |
TC2200007361.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200151829.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200151829.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200151827.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200151827.hg.1 |
NM_020243.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200151827.hg.1 |
TC2200007361.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200151827.hg.1 |
ENST00000216034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200151827.hg.1 |
NM_020243.4 |
Refseq |