Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00601216_g1
          See other KPNA4 GE Assays ›
          Gene Symbol:
          KPNA4
          Gene Name:
          karyopherin subunit alpha 4
          Gene Aliases:
          IPOA3, QIP1, SRP3
          Chromosome Location:
          Chr.3: 160494995 - 160565588 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00601216_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KPNA4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002268.4 NP_002259.1 3 - 4 502 149
          GenBank mRNA AB002533.2 - 3 - 4 205 149
          AK291041.1 - 3 - 4 454 149
          BC028691.2 - 3 - 4 447 4838709 149
          BC034493.2 - 3 - 4 502 4821602 149
          U93240.1 - 3 - 4 346 149

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KPNA4

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          NLS-bearing protein import into nucleus
          modulation by virus of host process
          response to hydrogen peroxide
          intracellular transport of virus
          protein binding
          nuclear localization sequence binding
          protein transporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000469804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000469804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000469804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000469804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000483437.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000483437.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000483437.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 ENST00000483437.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000334256.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000469804.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000469804.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000469804.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000469804.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000483437.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000483437.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000483437.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 ENST00000483437.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq
          Affymetrix Human Clariom D Assay TC0300012970.hg.1 NM_002268.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline