Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00606857_mH
          See other CELA2A GE Assays ›
          Gene Symbol:
          CELA2A
          Gene Name:
          chymotrypsin like elastase 2A
          Gene Aliases:
          AOMS4, ELA2A, PE-1
          Chromosome Location:
          Chr.1: 15456732 - 15472091 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00606857_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CELA2A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033440.3 NP_254275.1 6 - 7 661 133
          GenBank mRNA AK056678.1 - 1364 133
          AK312198.1 - 6 - 7 661 133
          BC007031.1 - 6 - 7 664 133
          BC069331.1 - 6 - 7 664 133
          BC069432.1 - 6 - 7 664 133
          BQ787677.1 - 2 - 3 270 133
          M16631.1 - 6 - 7 660 133
          M16652.1 - 6 - 7 665 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100018177.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01006265.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CELA2A

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          response to insulin
          regulation of insulin secretion
          regulation of platelet aggregation
          insulin catabolic process
          endopeptidase activity
          serine-type endopeptidase activity
          protein binding
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          serine hydrolase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01003100.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003100.hg.1 ENST00000375924.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003100.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01003100.hg.1 TC01006265.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01003098.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003098.hg.1 ENST00000375924.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003098.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01003098.hg.1 TC01006265.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01003101.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003101.hg.1 ENST00000375924.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003101.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01003101.hg.1 TC01006265.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000422901.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000422901.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000422901.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000422901.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000488764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000488764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000488764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000488764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 ENST00000488764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01003091.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003091.hg.1 ENST00000375924.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003091.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003091.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01003091.hg.1 TC01006265.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01003090.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003090.hg.1 ENST00000375924.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003090.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003090.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01003090.hg.1 TC01006265.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01003096.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003096.hg.1 ENST00000375924.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003096.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01003096.hg.1 TC01006265.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01003093.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003093.hg.1 ENST00000375924.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01003093.hg.1 NM_033440.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01003093.hg.1 TC01006265.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100237085.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237085.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237085.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237085.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237085.hg.1 TC0100018177.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100237080.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237080.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237080.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237080.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237080.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237080.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237080.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237080.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237080.hg.1 TC0100018177.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100237087.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237087.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237087.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237087.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237087.hg.1 TC0100018177.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018177.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000375910.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000422901.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000422901.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000422901.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000422901.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000488764.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000488764.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000488764.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000488764.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 ENST00000488764.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018178.hg.1 NM_015849.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237079.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237079.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237079.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237079.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237079.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237079.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237079.hg.1 ENST00000497590.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237079.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237079.hg.1 TC0100018177.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100237083.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237083.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237083.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237083.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237083.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237083.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237083.hg.1 TC0100018177.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100237082.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237082.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237082.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237082.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237082.hg.1 ENST00000459653.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237082.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237082.hg.1 TC0100018177.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100237086.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237086.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237086.hg.1 ENST00000359621.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100237086.hg.1 NM_033440.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100237086.hg.1 TC0100018177.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 112831 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline