Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00607779_s1
          See other MRGPRX4 GE Assays ›
          Gene Symbol:
          MRGPRX4
          Gene Name:
          MAS related GPR family member X4
          Gene Aliases:
          GPCR, MRGX4, SNSR6
          Chromosome Location:
          Chr.11: 18172837 - 18174280 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00607779_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MRGPRX4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_054032.3 NP_473373.2 1 1189 127
          GenBank mRNA BC095509.1 - 1 1199 30706615 127
          BC107099.1 - 1 769 40015896 127
          BC107100.1 - 1 769 40015904 127
          CN835944.1 - 1 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100006988.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000235.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MRGPRX4

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07671126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16424821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16424780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28070952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07671116 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07671121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28070933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07671124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28070960 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28070941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07671125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28070955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16424829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16424795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28070911 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P161983 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P161983 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 20655 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 57244 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02402878 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P161983 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P161983 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671112 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424794 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424821 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070918 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424780 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424821 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424780 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671112 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424794 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16424785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07671111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28070918 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P161983 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07671113 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07671117 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07671116 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07671114 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07671110 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07671112 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07671111 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07671115 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P161983 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671111 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16424795 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16424812 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671120 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671117 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671127 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671116 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671110 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671115 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16424842 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671119 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07671113 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_054032_at - -
          Affymetrix Human Gene 1.0 ST Array 7938747 - -
          Affymetrix Human Gene 1.1 ST Array 7938746 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553504_PM_at - -
          Affymetrix Human Genome U219 Array 11735850_PM_at - -
          Affymetrix Human Genome U219 Array 11735850_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 ENST00000529793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 ENST00000529793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 ENST00000618161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 ENST00000618161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 NM_054032.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 NM_054032.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 NM_054032.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006988.hg.1 NM_054032.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11003110.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11003110.hg.1 NM_054032.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11003110.hg.1 TC11000235.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100149191.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149191.hg.1 NM_054032.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100149191.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149191.hg.1 NM_054032.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100149191.hg.1 TC1100006988.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 ENST00000529793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 ENST00000529793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 ENST00000618161.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 ENST00000618161.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 NM_054032.3 Refseq
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 NM_054032.3 Refseq
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 NM_054032.3 Refseq
          Affymetrix Human Clariom D Assay TC1100006988.hg.1 NM_054032.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100149190.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149190.hg.1 NM_054032.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100149190.hg.1 ENST00000314254.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149190.hg.1 NM_054032.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100149190.hg.1 TC1100006988.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1553504_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553504_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_054032_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553504_at4 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046188 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046188 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 155352 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1722227 BC095509.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1722227 CN835944.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1722227 BC095509.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1722227 CN835944.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1722227 BC095509.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1722227 CN835944.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1722227 BC095509.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1722227 CN835944.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1722227 BC095509.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1722227 CN835944.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_16876454-S NM_054032.3 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_16876454-S BC095509.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_16876454-S BC107099.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_16876454-S BC107100.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_16876454-S CN835944.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_9949-S - -
          Illumina human-6 v1.0 expression beadchip 0001240059 - -
          Illumina humanRef-8 v1.0 expression beadchip 0001240059 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1722227 BC095509.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1722227 CN835944.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1722227 BC095509.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1722227 CN835944.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 2340452 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_9949 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1722227 BC095509.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1722227 CN835944.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1722227 BC095509.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1722227 CN835944.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1722227 BC095509.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1722227 CN835944.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1722227 BC095509.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1722227 CN835944.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1722227 BC095509.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1722227 CN835944.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline