Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00609976_m1
          See other NGFR GE Assays ›
          Gene Symbol:
          NGFR
          Gene Name:
          nerve growth factor receptor
          Gene Aliases:
          CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR
          Chromosome Location:
          Chr.17: 49495293 - 49515008 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00609976_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NGFR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002507.4 NP_002498.1 2 - 3 330 59
          GenBank mRNA AK303278.1 - 2 - 3 398 59
          AK313654.1 - 2 - 3 330 59
          BC050309.1 - 2 - 3 330 59
          DA115173.1 - 2 - 3 330 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008209.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000645.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Human NGF Pathway Human TNF Superfamily Pathway HUMAN APOPTOSIS THROUGH DEATH RECEPTORS HUMAN GUIDANCE CUES AND GROWTH CONE MOTILITY HUMAN P53 MEDIATED APOPTOSIS Inventoried Human_NGF_Pathway Human_TNF_Superfamily_Pathway Human_Guidance_Cues_and_Growth_Cone_Motility Human_Apoptosis_through_Death_Receptors Human_p53_Mediated_Apoptosis Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NGFR

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular glucose homeostasis
          hair follicle development
          intracellular protein transport
          apoptotic process
          signal transduction
          Rho protein signal transduction
          nervous system development
          axon guidance
          central nervous system development
          circadian rhythm
          fibroblast growth factor receptor signaling pathway
          regulation of gene expression
          detection of temperature stimulus
          nerve development
          cell differentiation
          negative regulation of cell migration
          hair follicle morphogenesis
          circadian regulation of gene expression
          dorsal aorta development
          negative regulation of fibroblast growth factor receptor signaling pathway
          regulation of cell proliferation
          odontogenesis of dentin-containing tooth
          positive regulation of odontogenesis of dentin-containing tooth
          glucose homeostasis
          positive regulation of apoptotic process
          skin development
          fibroblast proliferation
          positive regulation of fibroblast proliferation
          rhythmic process
          neuron apoptotic process
          negative regulation of hair follicle development
          extrinsic apoptotic signaling pathway
          presynaptic modulation of chemical synaptic transmission
          positive regulation of protein localization to nucleus
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          cellular response to beta-amyloid
          positive regulation of endothelial cell apoptotic process
          positive regulation of apoptotic signaling pathway
          beta-amyloid binding
          transmembrane signaling receptor activity
          death receptor activity
          protein binding
          calmodulin binding
          coreceptor activity
          small GTPase binding
          ubiquitin protein ligase binding
          signaling receptor activity
          neurotrophin binding
          nerve growth factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000172229.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000504201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000509200.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000509200.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 ENST00000509200.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008209.hg.1 NM_002507.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline