Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00611286_m1
          See other SYT2 GE Assays ›
          Gene Symbol:
          SYT2
          Gene Name:
          synaptotagmin 2
          Gene Aliases:
          CMS7, MYSPC, SytII
          Chromosome Location:
          Chr.1: 202590596 - 202710444 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00611286_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SYT2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001136504.1 NP_001129976.1 3 - 4 533 72
          NM_177402.4 NP_796376.2 3 - 4 523 72
          XM_017000310.1 XP_016855799.1 2 - 3 2290 72
          XM_017000313.1 XP_016855802.1 3 - 4 1197 72
          GenBank mRNA AK090672.1 - 3 - 4 517 72
          AK094430.1 - 3 - 4 533 72
          BC100814.1 - 3 - 4 388 40000996 72
          BC100815.1 - 3 - 4 388 40000997 72
          BC100817.1 - 3 - 4 388 40001000 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100016960.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003705.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SYT2

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          vesicle fusion
          regulation of calcium ion-dependent exocytosis
          synaptic vesicle endocytosis
          calcium ion-regulated exocytosis of neurotransmitter
          positive regulation of dendrite extension
          calcium ion binding
          protein binding
          calcium-dependent phospholipid binding
          syntaxin binding
          clathrin binding
          inositol 1,3,4,5 tetrakisphosphate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01056864.hg.1 ENST00000367267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01056864.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01056864.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01056864.hg.1 NM_177402.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01056864.hg.1 TC01003705.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay TC0100016960.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100228422.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100228422.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100228422.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100228422.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100228422.hg.1 ENST00000367267.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100228422.hg.1 ENST00000367268.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100228422.hg.1 NM_001136504.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100228422.hg.1 NM_177402.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100228422.hg.1 TC0100016960.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline