Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00708924_s1
          See other LINC02871 GE Assays ›
          Gene Symbol:
          LINC02871
          Gene Name:
          long intergenic non-protein coding RNA 2871
          Gene Aliases:
          C20orf187
          Chromosome Location:
          Chr.20: 11026544 - 11029366 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00708924_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC02871
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_146878.1 4 429 83
          GenBank mRNA BX281604.1 - 4 429 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P431430 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 18882 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 6837 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P431430 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 233598_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2000006708.hg.1 ENST00000378252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006708.hg.1 ENST00000378252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006708.hg.1 ENST00000378252.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006708.hg.1 ENST00000378252.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006708.hg.1 ENST00000378252.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006708.hg.1 ENST00000378252.1 Ensembl
          Human Genome U133 Plus 2.0 Array 233598_at - -
          Human Genome U133 Plus 2.0 Array 233598_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233598_at9 - -
          Human Genome U133 Set 233598_PM_at - -
          Human Genome U133 Set 233598_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 694686 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1863135 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_122546 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_122546 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1827984 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1827984 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline