| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007170.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007170.hg.1 |
ENST00000517298.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007170.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007170.hg.1 |
TC08002580.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007167.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007167.hg.1 |
TC08002580.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007168.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007168.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007168.hg.1 |
TC08002580.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007169.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007169.hg.1 |
ENST00000517298.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007169.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007169.hg.1 |
TC08002580.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800185689.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185689.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185689.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185689.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185689.hg.1 |
TC0800012323.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012323.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185690.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185690.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185690.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185690.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185690.hg.1 |
TC0800012323.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800185687.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185687.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185687.hg.1 |
TC0800012323.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800185688.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185688.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185688.hg.1 |
ENST00000321764.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185688.hg.1 |
NM_198584.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185688.hg.1 |
TC0800012323.hg.1 |
Clariom D TC |