Search
Search

MBLAC1| Gene Symbol: MBLAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203397.3 | NP_981942.1 | 2 | 979 | 120 | |
| XM_005250250.4 | XP_005250307.1 | 2 | 1048 | 120 | ||
| GenBank mRNA | AW449216.1 | - | 1 | 267 | 120 | |
| AW772344.1 | - | 1 | 244 | 120 | ||
| BC031288.1 | - | 2 | 1249 | 120 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646251 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646254 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646230 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25898753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646237 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25898797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646243 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646245 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25898770 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646247 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646250 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646259 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15994698 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646229 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646242 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646244 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25898767 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646253 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646232 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646263 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646233 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05646257 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25898798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15994697 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P221198 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P221198 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 32587 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 13977 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01765252 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P221198 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P221198 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646229 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25898753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25898771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646241 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646233 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25898770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25898753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646241 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646233 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25898771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05646229 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25898770 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P221198 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P05646228 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P05646229 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P05646230 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P05646234 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P05646233 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P05646239 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P05646237 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P05646232 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P05646233 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15994689 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15994690 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15994694 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15994688 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15994691 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15994692 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P05646231 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P05646235 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15994695 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15994687 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P05646230 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P05646236 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15994693 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P05646234 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P05646229 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P05646232 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P221198 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32081097 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P421984 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646202 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646232 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646256 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646203 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646233 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646237 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15994695 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15994698 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646229 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646260 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646210 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646250 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646205 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646261 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646201 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15994685 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646254 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646263 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646195 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646242 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646251 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646190 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646243 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646192 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15994696 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646212 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646239 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646230 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646234 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15994697 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15994693 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646249 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646199 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646258 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646209 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05646196 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_203397_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8134739 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8134738 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8134737 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1554170_PM_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11734372_at | - | - |
| Affymetrix Human Genome U219 Array | 11734372_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | ENST00000421390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | ENST00000421390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | ENST00000421390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | ENST00000421390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009198.hg.1 | ENST00000398075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009198.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009198.hg.1 | TC07000613.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009197.hg.1 | ENST00000398075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009197.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009197.hg.1 | TC07000613.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700159842.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700159842.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700159842.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700159842.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700159842.hg.1 | TC0700008525.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | ENST00000398075.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | ENST00000421390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | ENST00000421390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | ENST00000421390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | ENST00000421390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008525.hg.1 | NM_203397.2 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1554170_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1554170_a_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_203397_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1554170_a_at9 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034795 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034795 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 220354 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 223215 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1791438 | AW772344.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1791438 | BC031288.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1791438 | AW449216.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1791438 | AW772344.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1791438 | BC031288.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1791438 | AW449216.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1791438 | AW772344.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1791438 | BC031288.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1791438 | AW449216.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1791438 | AW772344.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1791438 | BC031288.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1791438 | AW449216.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1791438 | AW772344.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1791438 | BC031288.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1791438 | AW449216.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_5827-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1791438 | AW772344.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1791438 | BC031288.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1791438 | AW449216.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1791438 | AW772344.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1791438 | BC031288.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1791438 | AW449216.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 3060487 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_5827 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1791438 | AW772344.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1791438 | BC031288.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1791438 | AW449216.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1791438 | AW772344.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1791438 | BC031288.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1791438 | AW449216.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1791438 | AW772344.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1791438 | BC031288.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1791438 | AW449216.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1791438 | AW772344.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1791438 | BC031288.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1791438 | AW449216.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1791438 | AW772344.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1791438 | BC031288.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1791438 | NM_203397.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1791438 | AW449216.1 | human_transcriptome |