Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00758053_m1
          See other HAS1 GE Assays ›
          Gene Symbol:
          HAS1
          Gene Name:
          hyaluronan synthase 1
          Gene Aliases:
          HAS
          Chromosome Location:
          Chr.19: 51713112 - 51723992 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00758053_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HAS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001297436.1 NP_001284365.1 2 - 3 753 157
          NM_001523.3 NP_001514.2 2 - 3 756 157
          XM_011526884.2 XP_011525186.1 2 - 3 740 157
          GenBank mRNA AY916554.1 - 1 53 172
          AY916555.1 - 1 - 2 53 157
          AY916556.1 - 1 - 2 53 157
          BC035837.1 - 2 - 3 5589083 157
          BX371636.2 - 3 - 4 536 157
          D84424.1 - 1 - 2 740 157
          U59269.1 - 2 - 3 732 157

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900011270.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19001786.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HAS1

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycosaminoglycan biosynthetic process
          cell adhesion
          negative regulation of fibroblast migration
          hyaluronan biosynthetic process
          cellular response to platelet-derived growth factor stimulus
          estrous cycle
          extracellular polysaccharide biosynthetic process
          extracellular matrix assembly
          hyaluronan synthase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19024269.hg.1 ENST00000222115.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19024269.hg.1 ENST00000376737.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19024269.hg.1 ENST00000376738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19024269.hg.1 ENST00000540069.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19024269.hg.1 NM_001523.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19024269.hg.1 TC19001786.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011270.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 TC1900011270.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900194906.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 TC1900011270.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900194904.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 NM_001523.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 TC1900011270.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 ENST00000222115.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 ENST00000540069.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 ENST00000594621.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 ENST00000601667.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 ENST00000601714.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 NM_001297436.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900194905.hg.1 NM_001523.3 Refseq

          Back To Top
           

          Citations (1)

          Glucosamine increases hyaluronic acid production in human osteoarthritic synovium explants.
          Authors
          Uitterlinden EJ, Koevoet JL, Verkoelen CF, Bierma-Zeinstra SM, Jahr H, Weinans H, Verhaar JA, van Osch GJ
          Journal
          Bmc Musculoskelet Disord - Page No 120 (Tue Jan 01 00:00:00 UTC 2008)
          Volume (Year)
          9 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline