| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006381.hg.1 |
ENST00000356464.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006381.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006381.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006381.hg.1 |
TC12000500.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006380.hg.1 |
ENST00000356464.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006380.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006380.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006380.hg.1 |
TC12000500.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006382.hg.1 |
ENST00000356464.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006382.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12006382.hg.1 |
TC12000500.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000548590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000548590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000548590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000548590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000548590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200156478.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200156478.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200156478.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200156478.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200156478.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200156478.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200156478.hg.1 |
TC1200007823.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200156479.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200156479.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200156479.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200156479.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200156479.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200156479.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200156479.hg.1 |
TC1200007823.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000548590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000548590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000548590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000548590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000548590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
ENST00000552361.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007823.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200156480.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200156480.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200156480.hg.1 |
ENST00000356464.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200156480.hg.1 |
NM_001029.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200156480.hg.1 |
TC1200007823.hg.1 |
Clariom D TC |