| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510121.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510121.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510121.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510121.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014747.hg.1 |
ENST00000326877.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014747.hg.1 |
ENST00000382224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014747.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014747.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014747.hg.1 |
ENST00000539056.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014747.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014747.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014747.hg.1 |
TC04001070.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014749.hg.1 |
ENST00000326877.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014749.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014749.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014749.hg.1 |
ENST00000539056.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014749.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014749.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014749.hg.1 |
TC04001070.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014752.hg.1 |
ENST00000326877.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014752.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014752.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014752.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014752.hg.1 |
TC04001070.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169727.hg.1 |
TC0400010168.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169729.hg.1 |
TC0400010168.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400169733.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169733.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169733.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169733.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169733.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169733.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400169733.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169733.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400169733.hg.1 |
TC0400010168.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000326877.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382224.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000382226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510121.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510121.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510121.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510121.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000510451.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
ENST00000512376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_001166139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010168.hg.1 |
NM_153686.8 |
Refseq |