Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00818121_m1
          See other FOXO3 GE Assays ›
          Gene Symbol:
          FOXO3
          Gene Name:
          forkhead box O3
          Gene Aliases:
          AF6q21, FKHRL1, FKHRL1P2, FOXO2, FOXO3A
          Chromosome Location:
          Chr.6: 108559825 - 108684774 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00818121_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FOXO3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001455.4 NP_001446.1 1 - 2 916 214
          NM_201559.3 NP_963853.1 2 - 3 938 214
          GenBank mRNA AF032886.1 - 1 - 2 1548 214
          AK122861.1 - 2 - 3 523 214
          BC020227.1 - 2 - 3 928 214
          BC021224.2 - 2 - 3 928 214
          BC068552.1 - 1 - 2 967 214

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600009059.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000868.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Human Androgen Panel HUMAN INSULIN RECEPTOR PATHWAY HUMAN PI3K SIGNALING IN B-LYMPHOCYTE HUMAN IGF1R SIGNALING Inventoried Human_Androgen Human_PI3K_Signaling_in_B-Lymphocyte Human_IGF1R_Signaling Human_Insulin_Receptor_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FOXO3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ovulation from ovarian follicle
          initiation of primordial ovarian follicle growth
          antral ovarian follicle growth
          oocyte maturation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from mitochondrial promoter
          regulation of translation
          apoptotic process
          response to xenobiotic stimulus
          positive regulation of autophagy
          positive regulation of muscle atrophy
          DNA damage response, signal transduction by p53 class mediator
          negative regulation of cell migration
          response to nutrient levels
          tumor necrosis factor-mediated signaling pathway
          cellular response to oxidative stress
          cellular response to glucose starvation
          glucose homeostasis
          response to starvation
          positive regulation of apoptotic process
          positive regulation of neuron apoptotic process
          positive regulation of regulatory T cell differentiation
          positive regulation of erythrocyte differentiation
          negative regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          brain morphogenesis
          canonical Wnt signaling pathway
          response to fatty acid
          cellular response to glucose stimulus
          cellular response to corticosterone stimulus
          cellular response to hypoxia
          response to dexamethasone
          negative regulation of canonical Wnt signaling pathway
          neuronal stem cell population maintenance
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of hydrogen peroxide-mediated programmed cell death
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          positive regulation of reactive oxygen species biosynthetic process
          cellular response to beta-amyloid
          cellular response to nerve growth factor stimulus
          response to water-immersion restraint stress
          regulation of neural precursor cell proliferation
          positive regulation of endothelial cell apoptotic process
          regulation of reactive oxygen species metabolic process
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription cofactor binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          beta-catenin binding
          protein kinase binding
          chromatin DNA binding
          mitochondrial RNA polymerase binding promoter specificity activity
          sequence-specific DNA binding
          sequence-specific double-stranded DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06010146.hg.1 ENST00000343882.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06010146.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06010146.hg.1 ENST00000540258.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06010146.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06010146.hg.1 NM_001455.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06010146.hg.1 NM_201559.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06010146.hg.1 TC06000868.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 NM_001455.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 NM_201559.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 NM_001455.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 NM_201559.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600162211.hg.1 TC0600009059.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000343882.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000406360.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 ENST00000540898.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_001455.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_201559.2 Refseq
          Affymetrix Human Clariom D Assay TC0600009059.hg.1 NM_201559.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline