| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000299440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000299440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000299440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000299440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000529126.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000529126.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000529126.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
NM_000448.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
NM_000448.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
NM_000448.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007306.hg.1 |
NM_000448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000299440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000299440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000299440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000299440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000529126.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000529126.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000529126.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
NM_000448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
NM_000448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
NM_000448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007306.hg.1 |
NM_000448.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151566.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151566.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151566.hg.1 |
TC1100007306.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151567.hg.1 |
ENST00000529126.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151567.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151567.hg.1 |
ENST00000529126.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151567.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151567.hg.1 |
TC1100007306.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151582.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151582.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151582.hg.1 |
TC1100007306.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151583.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151583.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151583.hg.1 |
TC1100007306.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151573.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151573.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151573.hg.1 |
TC1100007306.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151574.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151574.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151574.hg.1 |
TC1100007306.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151572.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151572.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151572.hg.1 |
TC1100007306.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151569.hg.1 |
ENST00000529126.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151569.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151569.hg.1 |
ENST00000529126.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151569.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151569.hg.1 |
TC1100007306.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151571.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151571.hg.1 |
ENST00000534663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151571.hg.1 |
TC1100007306.hg.1 |
Clariom D TC |