| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061438.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061438.hg.1 |
TC01004073.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000420469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000420469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000420469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018101.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061436.hg.1 |
ENST00000420469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061436.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061436.hg.1 |
TC01004073.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061442.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061442.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061442.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01061442.hg.1 |
TC01004073.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100236231.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236231.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236231.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236231.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236231.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236231.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236231.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236231.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236231.hg.1 |
TC0100018101.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000366487.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000420469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000420469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000420469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
ENST00000617752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NM_198074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018101.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236226.hg.1 |
ENST00000420469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236226.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236226.hg.1 |
ENST00000420469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236226.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236226.hg.1 |
TC0100018101.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100236228.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236228.hg.1 |
NR_027309.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236228.hg.1 |
TC0100018101.hg.1 |
Clariom D TC |