Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00859907_m1
          See other SYT7 GE Assays ›
          Gene Symbol:
          SYT7
          Gene Name:
          synaptotagmin 7
          Gene Aliases:
          IPCA-7, IPCA7, PCANAP7, SYT-VII, SYTVII
          Chromosome Location:
          Chr.11: 61513716 - 61588404 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00859907_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SYT7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252065.1 NP_001238994.1 5 - 6 630 81
          NM_001300773.1 NP_001287702.1 5 - 6 537 81
          NM_004200.3 NP_004191.2 4 - 5 405 81
          XM_005274383.4 XP_005274440.1 8 - 9 1292 81
          XM_005274384.2 XP_005274441.1 7 - 8 1296 81
          XM_005274385.4 XP_005274442.1 7 - 8 1062 81
          XM_005274387.4 XP_005274444.1 2 - 3 1329 81
          XM_011545335.2 XP_011543637.1 8 - 9 1258 81
          XM_011545336.2 XP_011543638.1 7 - 8 1133 81
          XM_011545337.2 XP_011543639.1 7 - 8 1125 81
          XM_011545338.2 XP_011543640.1 7 - 8 1030 81
          XM_011545339.2 XP_011543641.1 6 - 7 757 81
          XM_011545340.2 XP_011543642.1 5 - 6 623 81
          XM_011545341.2 XP_011543643.1 5 - 6 527 81
          XM_011545343.2 XP_011543645.1 4 - 5 391 81
          GenBank mRNA AF038535.1 - 4 - 5 398 81
          BC125170.1 - 4 - 5 363 40121712 81
          BE644642.1 - 4 - 5 405 81
          BM679451.1 - 3 - 4 81
          CA868518.1 - 5 - 6 503 81

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SYT7

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane repair
          vesicle fusion
          phagocytosis
          regulation of calcium ion-dependent exocytosis
          synaptic vesicle recycling
          positive regulation of calcium ion-dependent exocytosis
          regulation of bone remodeling
          calcium ion-regulated exocytosis of neurotransmitter
          regulation of phagocytosis
          regulation of insulin secretion
          regulation of glucagon secretion
          vesicle-mediated cholesterol transport
          phagosome-lysosome fusion
          short-term synaptic potentiation
          calcium ion regulated lysosome exocytosis
          calcium ion binding
          protein binding
          calmodulin binding
          calcium-dependent phospholipid binding
          phosphatidylinositol-4,5-bisphosphate binding
          syntaxin binding
          clathrin binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline