| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300158702.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300158702.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300158702.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300158702.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300158702.hg.1 |
TC0300008269.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300158705.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300158705.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300158705.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300158705.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300158705.hg.1 |
TC0300008269.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008269.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300158707.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300158707.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300158707.hg.1 |
ENST00000609293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300158707.hg.1 |
NR_110039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300158707.hg.1 |
TC0300008269.hg.1 |
Clariom D TC |