| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165960 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366745 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366790 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366797 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165925 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165946 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165938 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165914 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165910 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165945 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366756 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165932 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165953 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366774 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165936 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15873446 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366768 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165935 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165954 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165948 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366740 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165956 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165921 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165928 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366758 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165922 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165958 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165937 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165906 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366658 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165927 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165923 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165940 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165902 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366750 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165941 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165929 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165951 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366746 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165911 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366754 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165917 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165905 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165955 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366694 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
124293 |
AF468496.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
124293 |
AF468497.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
124293 |
AK095391.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
124293 |
AK123614.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
124293 |
AY188947.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
124293 |
AY188949.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
124293 |
BC132971.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
124293 |
BC132973.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
124293 |
NM_133263.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
105169 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
290358 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_3022 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P145485 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165921 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165940 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165909 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366740 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165955 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366768 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165936 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165946 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165917 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165907 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366756 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165910 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165927 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366754 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873457 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165948 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165922 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165925 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165914 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165919 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165938 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366797 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366790 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165951 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366758 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165935 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366784 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366774 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165953 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165923 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165928 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165956 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366791 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165943 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873446 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165945 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165929 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165932 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366750 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165958 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165943 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165940 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366754 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165921 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366790 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165948 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366784 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165910 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366758 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366774 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165951 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165953 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873457 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165917 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366750 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873446 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165938 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165955 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165925 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165907 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165922 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165945 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165946 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165935 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165928 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165927 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165958 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366740 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165919 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366791 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165929 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165909 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165932 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366768 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165923 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165936 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366756 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165914 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165956 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366797 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165912 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165908 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165938 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165927 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165928 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165918 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165916 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165955 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165917 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165920 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165923 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165933 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165951 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165953 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165932 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165948 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165945 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165922 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165925 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165935 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165910 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165936 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165914 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165946 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165907 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165941 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05165950 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15873333 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15873336 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15873337 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15873478 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15873332 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15873335 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P05165882 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15873479 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P05165883 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P31905685 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165923 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165948 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165904 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165925 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165957 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15873455 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165946 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165938 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165945 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165916 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165912 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165958 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15873448 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165960 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165935 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165910 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165917 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165920 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165953 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165936 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165922 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15873457 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165914 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165955 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165932 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165903 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165933 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165950 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165927 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165941 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165951 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165928 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15873453 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165907 |
- |
- |