Search
Search
| Gene Symbol: RYR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035.2 | NP_001026.2 | 89 - 90 | 12078 | 73 | |
| XM_006711802.3 | XP_006711865.1 | 91 - 92 | 12326 | 73 | ||
| XM_006711803.3 | XP_006711866.1 | 91 - 92 | 12323 | 73 | ||
| XM_006711804.3 | XP_006711867.1 | 90 - 91 | 12302 | 73 | ||
| XM_006711805.3 | XP_006711868.1 | 90 - 91 | 12296 | 73 | ||
| XM_006711806.3 | XP_006711869.1 | 90 - 91 | 12290 | 73 | ||
| XM_006711807.3 | XP_006711870.1 | 89 - 90 | 12266 | 73 | ||
| XM_006711808.3 | XP_006711871.1 | 88 - 89 | 12089 | 73 | ||
| XM_006711810.3 | XP_006711873.1 | 88 - 89 | 12233 | 73 | ||
| XM_017002028.1 | XP_016857517.1 | 90 - 91 | 12305 | 73 | ||
| GenBank mRNA | AK294792.1 | - | 29 - 30 | 3153 | 73 | |
| X98330.1 | - | 89 - 90 | 12079 | 73 | ||
Set Membership: |
|