| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000581328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000581328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000581328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
ENST00000581328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007585.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007585.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007585.hg.1 |
TC17000613.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700159907.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700159907.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700159907.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700159907.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700159907.hg.1 |
TC1700008135.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000177694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000581328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000581328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000581328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
ENST00000581328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008135.hg.1 |
NM_013351.1 |
Refseq |