| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03007339.hg.1 |
ENST00000345716.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03007339.hg.1 |
ENST00000414197.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03007339.hg.1 |
ENST00000433196.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03007339.hg.1 |
ENST00000479054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03007339.hg.1 |
NM_007184.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03007339.hg.1 |
TC03000335.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
ENST00000345716.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
ENST00000467594.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
ENST00000479054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
ENST00000489895.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
NM_007184.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
ENST00000345716.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
ENST00000467594.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
ENST00000479054.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
ENST00000489895.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
NM_007184.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300153935.hg.1 |
TC0300007482.hg.1 |
Clariom D TC |