| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044406.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044406.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044406.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044406.hg.1 |
TC01006361.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044407.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044407.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044407.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044407.hg.1 |
TC01006361.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100242311.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242311.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242311.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242311.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242311.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242311.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242311.hg.1 |
TC0100018451.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100242312.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242312.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242312.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242312.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242312.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242312.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242312.hg.1 |
TC0100018451.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000370466.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000463660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
ENST00000493802.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018451.hg.1 |
NM_004120.4 |
Refseq |