| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035414.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035414.hg.1 |
ENST00000470487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035414.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035414.hg.1 |
TC03001978.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035416.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035416.hg.1 |
ENST00000470487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035416.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035416.hg.1 |
TC03001978.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035418.hg.1 |
ENST00000470487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035418.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035418.hg.1 |
TC03001978.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035413.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035413.hg.1 |
ENST00000470487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035413.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035413.hg.1 |
TC03001978.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035415.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035415.hg.1 |
ENST00000470487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035415.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035415.hg.1 |
TC03001978.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035417.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035417.hg.1 |
ENST00000470487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035417.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035417.hg.1 |
TC03001978.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300197652.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197652.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197652.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197652.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197652.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197652.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197652.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197652.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197652.hg.1 |
TC0300013058.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300197654.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197654.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197654.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197654.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197654.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197654.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197654.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197654.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197654.hg.1 |
TC0300013058.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300197655.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197655.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197655.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197655.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197655.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197655.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197655.hg.1 |
TC0300013058.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
ENST00000479415.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013058.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197653.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197653.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197653.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197653.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197653.hg.1 |
ENST00000309027.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197653.hg.1 |
ENST00000470487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300197653.hg.1 |
NM_001308155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197653.hg.1 |
NM_014498.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300197653.hg.1 |
TC0300013058.hg.1 |
Clariom D TC |