Search
Search

UBOX5| Gene Symbol: UBOX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014948.3 | NP_055763.1 | 4 - 5 | 1610 | 88 | |
| GenBank mRNA | AB020667.1 | - | 4 - 5 | 1578 | 88 | |
| AK222876.1 | - | 4 - 5 | 1535 | 88 | ||
| BC000515.2 | - | 4 - 5 | 1530 | 2822816 | 88 | |
| CR457089.1 | - | 3 - 4 | 1424 | 88 | ||
Set Membership: |
|